missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-tert-butylanisole 98.0+%, TCI America™

Catalog No. B0723500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0723500G 500 g
B072325G 25 g
B0723100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0723500G Supplier TCI America Supplier No. B0723500G
Only null left

Chemical Identifiers

CAS 121-00-6
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.247
MDL Number MFCD00040484
InChI Key MRBKEAMVRSLQPH-UHFFFAOYSA-N
Synonym 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy
PubChem CID 8456
ChEBI CHEBI:76358
IUPAC Name 2-tert-butyl-4-methoxyphenol
SMILES CC(C)(C)C1=C(C=CC(=C1)OC)O

Specifications

Melting Point 60°C
Quantity 500 g
UN Number 3077
Formula Weight 180.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-tert-butylanisole
EINECSNumber (3)-0608&(9)-1199&(9)-1532
RTECSNumber SK1575000
TSCA Yes