missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxycinnamic acid, Mixture of isomers, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012635
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012635 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012635 Supplier Merck Emd Millipore Supplier No. 90034100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

4-Hydroxy-3-methoxycinnamic acid is a ubiquitous plant constituent primarily occurring in seeds and leaves. It arises from the metabolism of phenylalanine and tyrosine.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Chemical Name or Material Analytical Standard
Linear Formula C10H10O4
Quantity 100 mg
MDL Number MFCD00004400
Synonym Ferulic acid
Grade Analytical Standard
Physical Form Neat