missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxybenzonitrile 98.0+%, TCI America™

Catalog No. H06205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06205G 5 g
H062025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06205G Supplier TCI America Supplier No. H06205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4421-08-3
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
MDL Number MFCD00001820
InChI Key QJRWLNLUIAJTAD-UHFFFAOYSA-N
Synonym vanillonitrile, m-anisonitrile, 4-hydroxy, 4-hydroxy-3-methoxy-benzonitrile, 3-methoxy-4-hydroxybenzonitrile, 4-hydroxy-3-methoxybenzenecarbonitrile, pubchem13112, 2-methoxy-4-cyanophenol, 4-cyano-2-methoxyphenol, 5-cyano-2-hydroxyanisole, 2-methoxy-4-cyano-phenol
PubChem CID 78135
IUPAC Name 4-hydroxy-3-methoxybenzonitrile
SMILES COC1=C(C=CC(=C1)C#N)O

Specifications

Melting Point 87°C
Quantity 5 g
UN Number 3439
Formula Weight 149.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-methoxybenzonitrile