missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dimethylbenzonitrile 95.0+%, TCI America™

Catalog No. D27905G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27905G 5 g
D27901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27905G Supplier TCI America Supplier No. D27905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4198-90-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00221716
InChI Key WFYGXOWFEIOHCZ-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-hydroxybenzonitrile, 4-cyano-2,6-dimethylphenol, 2,6-dimethyl-4-cyanophenol, 4-hydroxy-3,5-dimethyl-benzonitrile, 3,5-dimethyl-4-hydroxy benzonitrile, benzonitrile, 3,5-dimethyl-4-hydroxy, 4-hydroxy-3,5-dimethylbenzenecarbonitrile, benzonitrile, 4-hydroxy-3,5-dimethyl, pubchem11065, acmc-209jmm
PubChem CID 20176
IUPAC Name 4-hydroxy-3,5-dimethylbenzonitrile
SMILES CC1=CC(=CC(=C1O)C)C#N

Specifications

Melting Point 125°C
Quantity 5 g
Formula Weight 147.18
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3,5-dimethylbenzonitrile
RTECSNumber DI4359000
TSCA No