missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dimethylbenzaldehyde 98.0+%, TCI America™

Catalog No. H10235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10235G 5 g
H102325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10235G Supplier TCI America Supplier No. H10235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2233-18-3
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00006946
InChI Key UYGBSRJODQHNLQ-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-hydroxybenzaldehyde, benzaldehyde, 4-hydroxy-3,5-dimethyl, 4-formyl-2,6-xylenol, 4-hydroxy-3,5-dimethyl-benzaldehyde, unii-23pa2pmp9f, 23pa2pmp9f, syringnldehyde, zlchem 510, pubchem8252, buttpark 9657-68
PubChem CID 75222
IUPAC Name 4-hydroxy-3,5-dimethylbenzaldehyde
SMILES CC1=CC(C=O)=CC(C)=C1O

Specifications

Melting Point 117°C
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3,5-dimethylbenzaldehyde