missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. H14951G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H14951G Supplier TCI America Supplier No. H14951G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-Hydroxy-2-methylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 493035-82-8
Melting Point 104°C
Color White-Yellow
Molecular Formula C7H9BO3
Quantity 1 g
MDL Number MFCD03788424
Synonym 4-hydroxy-2-methylbenzeneboronic acid, 4-hydroxy-2-methyl phenylboronic acid, 4-borono-3-methylphenol, 4-hydroxy-2-methylphenyl boronic acid, 4-hydroxy-2-methyl phenyl boronic acid, 2-borono-5-hydroxytoluene, boronic acid, 4-hydroxy-2-methylphenyl, 5-hydroxytoluene-2-boronic acid, 4-hydroxy-2-methylphenylboronicacid, tavaborole impurity 3
InChI Key OYIYNIONWDBJIF-UHFFFAOYSA-N
SMILES CC1=CC(O)=CC=C1B(O)O
IUPAC Name (4-hydroxy-2-methylphenyl)boronic acid
Molecular Weight (g/mol) 151.96
PubChem CID 2773448
Formula Weight 151.96
Physical Form Crystalline Powder
Show More Show Less