missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2,6-dimethoxybenzaldehyde 97.0+%, TCI America™

Catalog No. H13261G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13261G 1 g
H13265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13261G Supplier TCI America Supplier No. H13261G

Chemical Identifiers

CAS 22080-96-2
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD01407645
InChI Key HZWPJAZIRZFCGX-UHFFFAOYSA-N
PubChem CID 529894
IUPAC Name 4-hydroxy-2,6-dimethoxybenzaldehyde
SMILES COC1=CC(O)=CC(OC)=C1C=O

Specifications

Melting Point 220°C
Quantity 1 g
Formula Weight 182.18
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2,6-dimethoxybenzaldehyde