missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2(5H)-furanone, 95%

Catalog No. AC235260050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC235260050 5 g Glass bottle
AC235260010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC235260050 Supplier Thermo Scientific Chemicals Supplier No. 235260050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 541-57-1 | C4H4O3 | 100.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 541-57-1
Molecular Formula C4H4O3
Molecular Weight (g/mol) 100.07
MDL Number MFCD00052187
InChI Key JZQBAGOECGRTSA-UHFFFAOYSA-N
Synonym 4-hydroxyfuran-2 5h-one, 4-hydroxy-2,5-dihydrofuran-2-one, 4-hydroxy-2 5h-furanone, 2 5h-furanone, 4-hydroxy, 4-hydroxy-5h-furan-2-one, 5-hydroxyfuran-3-one, 5-hydroxyfuran-3 2h-one, pubchem21770, acmc-209xna, tetronic acid 5g
PubChem CID 54683813
IUPAC Name 3-hydroxy-2H-furan-5-one
SMILES OC1=CC(=O)OC1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 135°C to 137°C
Color Beige-Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,acetonitrile,dmso
Formula Weight 100.07
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2(5H)-furanone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
EINECSNumber 208-785-3

RUO – Research Use Only