missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Benzoate Free Radical 97.0+%, TCI America™

Catalog No. p-7130778
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08781G 1 g
H08785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08781G Supplier TCI America Supplier No. H08781G

Chemical Identifiers

CAS 3225-26-1
Molecular Formula C16H22NO3
Molecular Weight (g/mol) 276.356
MDL Number MFCD00075563
InChI Key MJEDTBDGYVATPI-UHFFFAOYSA-N
Synonym 4-Hydroxy-TEMPO Benzoate, 4-Benzoyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Benzoyloxy-TEMPO
PubChem CID 2847363
IUPAC Name (1-$l^{1}-oxidanyl-2,2,6,6-tetramethylpiperidin-4-yl) benzoate
SMILES CC1(CC(CC(N1[O])(C)C)OC(=O)C2=CC=CC=C2)C

Specifications

Melting Point 104°C
Quantity 1 g
Formula Weight 276.36
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Benzoate Free Radical [Catalyst for Oxidation]