missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98%

Catalog No. AAA1697414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1697414 25 g
AAA1697406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1697414 Supplier Thermo Scientific Chemicals Supplier No. A1697414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 315-30-0 | C5H4N4O | 136.11 g/mol

Xanthine oxidase inhibitor; decreases uric acid production

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 315-30-0
Molecular Formula C5H4N4O
Molecular Weight (g/mol) 136.11
MDL Number MFCD00599413
InChI Key OFCNXPDARWKPPY-UHFFFAOYSA-N
Synonym allopurinol, 1h-pyrazolo 3,4-d pyrimidin-4-ol, zyloprim, zyloric, lopurin, atisuril, bleminol, caplenal, uripurinol, embarin
PubChem CID 2094
ChEBI CHEBI:40279
IUPAC Name 1,2-dihydropyrazolo[3,4-d]pyrimidin-4-one
SMILES O=C1N=CN=C2NNC=C12

Specifications

Melting Point >300°C
Odor Odorless
Quantity 25 g
UN Number UN2811
Formula Weight 136.11
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine
Hazard Category H301-H317-H500
Hazard Statement GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P310-P302+P352-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 206-250-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only