missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidine 98.0+%, TCI America™

Numéro de catalogue. H09315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H09315G 5 g
H093125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. H09315G Fournisseur TCI America Code fournisseur H09315G
Il en reste null

Identifiants chimiques

CAS 52722-86-8
Molecular Formula C11H23NO2
Molecular Weight (g/mol) 201.31
MDL Number MFCD01321174
InChI Key STEYNUVPFMIUOY-UHFFFAOYSA-N
Synonym 1-(2-Hydroxyethyl)-2,2,6,6-tetramethyl-4-piperidinol, 4-Hydroxy-2,2,6,6-tetramethyl-1-piperidineethanol
PubChem CID 104301
IUPAC Name 1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidin-4-ol
SMILES CC1(CC(CC(N1CCO)(C)C)O)C

Spécifications

Melting Point 186°C
Color White
Quantity 5 g
Formula Weight 201.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidine