missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzenesulfonic Acid 98.0+%, TCI America™

Catalog No. H1662500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1662500G 500 g
H166225G 25 g
H1662100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H1662500G Supplier TCI America Supplier No. H1662500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-71-5
Molecular Formula C6H8N2O3S
Molecular Weight (g/mol) 188.20
MDL Number MFCD00025097
InChI Key IOMZCWUHFGMSEJ-UHFFFAOYSA-N
Synonym 4-hydrazinobenzenesulfonic acid, phenylhydrazine-4-sulfonic acid, benzenesulfonic acid, 4-hydrazino, p-hydrazinobenzenesulfonic acid, p-sulfophenylhydrazine, 4-sulfophenylhydrazine, 4-hydrazinobenzenesulphonic acid, phenylhydrazinesulfonic acid, phenylhydrazine-p-sulfonic acid, sulfanilic acid, n-amino
PubChem CID 66825
IUPAC Name 4-hydrazinylbenzene-1-sulfonic acid
SMILES NNC1=CC=C(C=C1)S(O)(=O)=O

Specifications

Melting Point 286°C
Color White-Yellow
Quantity 500 g
Formula Weight 188.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydrazinobenzenesulfonic Acid
EINECSNumber (3)-1997
RTECSNumber DB6900000
TSCA Yes