missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzenesulfonamide Hydrochloride 98.0+%, TCI America™

Catalog No. p-7130929
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12305G 5 g
H123025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12305G Supplier TCI America Supplier No. H12305G

Chemical Identifiers

CAS 17852-52-7
Molecular Formula C6H10ClN3O2S
Molecular Weight (g/mol) 223.675
MDL Number MFCD00052262
InChI Key IKEURONJLPUALY-UHFFFAOYSA-N
Synonym 4-hydrazinylbenzenesulfonamide hydrochloride, 4-hydrazinobenzene-1-sulfonamide hydrochloride, 4-hydrazinobenzenesulfonamide hydrochloride, 4-sulfonamidephenylhydrazine hcl, 4-aminosulfonylphenylhydrazine hydrochloride, 4-hydrazinylbenzenesulfonamide hydrochloride 1:x, 4-sulfamoylphenyl hydrazine hydrochloride, 4-sulfonamide-phenylhydrazine hcl, phenylhydrazine-4-sulfonamide hydrochloride
PubChem CID 2794567
IUPAC Name 4-hydrazinylbenzenesulfonamide;hydrochloride
SMILES C1=CC(=CC=C1NN)S(=O)(=O)N.Cl

Specifications

Melting Point 211°C
Quantity 5 g
Formula Weight 223.68
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydrazinobenzenesulfonamide Hydrochloride