missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydrazinobenzenesulfonamide Hydrochloride 98.0+%, TCI America™
Supplier: TCI America H123025G
Specifications
| 4-Hydrazinobenzenesulfonamide Hydrochloride | |
| 211°C | |
| MFCD00052262 | |
| 4-hydrazinylbenzenesulfonamide hydrochloride, 4-hydrazinobenzene-1-sulfonamide hydrochloride, 4-hydrazinobenzenesulfonamide hydrochloride, 4-sulfonamidephenylhydrazine hcl, 4-aminosulfonylphenylhydrazine hydrochloride, 4-hydrazinylbenzenesulfonamide hydrochloride 1:x, 4-sulfamoylphenyl hydrazine hydrochloride, 4-sulfonamide-phenylhydrazine hcl, phenylhydrazine-4-sulfonamide hydrochloride | |
| C1=CC(=CC=C1NN)S(=O)(=O)N.Cl | |
| 223.675 | |
| 223.68 | |
| Crystalline Powder |
| 17852-52-7 | |
| C6H10ClN3O2S | |
| 25 g | |
| IKEURONJLPUALY-UHFFFAOYSA-N | |
| 4-hydrazinylbenzenesulfonamide;hydrochloride | |
| 2794567 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 17852-52-7 | |
| 223.675 | |
| IKEURONJLPUALY-UHFFFAOYSA-N | |
| 2794567 | |
| C1=CC(=CC=C1NN)S(=O)(=O)N.Cl |
| C6H10ClN3O2S | |
| MFCD00052262 | |
| 4-hydrazinylbenzenesulfonamide hydrochloride, 4-hydrazinobenzene-1-sulfonamide hydrochloride, 4-hydrazinobenzenesulfonamide hydrochloride, 4-sulfonamidephenylhydrazine hcl, 4-aminosulfonylphenylhydrazine hydrochloride, 4-hydrazinylbenzenesulfonamide hydrochloride 1:x, 4-sulfamoylphenyl hydrazine hydrochloride, 4-sulfonamide-phenylhydrazine hcl, phenylhydrazine-4-sulfonamide hydrochloride | |
| 4-hydrazinylbenzenesulfonamide;hydrochloride |
Safety and Handling
TSCA : No