accessibility menu, dialog, popup

UserName

HClH2NNHSNH2OO

CAS RN 17852-52-7

IUPAC Name: hydrogen 4-hydrazinylbenzene-1-sulfonamide chloride
Synonyms: hydrogen 4-hydrazinylbenzenesulfonamide chloride
Molecular Weight (g/mol): 223.68
Molecular Formula: C6H10ClN3O2S
InChi Key: IKEURONJLPUALY-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=C(C=C1)S(N)(=O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)