missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hexylphenol 98.0+%, TCI America™
Spécifications
Spécifications
| Color | White-Yellow |
| Quantity | 5 g |
| UN Number | 2430 |
| Formula Weight | 178.28 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 4-Hexylphenol |
Sécurité et manipulation
| TSCA | Yes |