missing translation for 'onlineSavingsMsg'
Learn More

4-Formylphenyl beta-D-Allopyranoside 98.0+%, TCI America™

Catalog No. F05425G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05425G Supplier TCI America Supplier No. F05425G

Chemical Identifiers

CAS 80154-34-3
Molecular Formula C13H16O7
Molecular Weight (g/mol) 284.264
InChI Key OLZAGZCCJJBKNZ-SYLRKERUSA-N
Synonym helicide, helicid, hilicidum, 4-formylphenyl beta-d-allopyranoside, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy benzaldehyde, 4-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy benzaldehyde, 4-formylphenyl ss-d-allopyranoside, 4-beta-d-allopyranosyloxy benzaldehyde, 4-formylphenyl b-d-allopyranoside
PubChem CID 12896796
IUPAC Name 4-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzaldehyde
SMILES C1=CC(=CC=C1C=O)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 190°C
Color White
Quantity 5 g
Formula Weight 284.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Formylphenyl beta-D-Allopyranoside