missing translation for 'onlineSavingsMsg'
Learn More

4'-Formylbenzo-15-crown 5-Ether 98.0+%, TCI America™

Numéro de catalogue. F04481G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
F04481G 1 g
F0448100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. F04481G Fournisseur TCI America Code fournisseur F04481G
Il en reste null

Identifiants chimiques

CAS 60835-73-6
Molecular Formula C15H20O6
Molecular Weight (g/mol) 296.319
MDL Number MFCD00192180
InChI Key MBJIKIAWNPEHOR-UHFFFAOYSA-N
Synonym 2,3-(4-Formylbenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene
PubChem CID 324243
IUPAC Name 2,5,8,11,14-pentaoxabicyclo[13.4.0]nonadeca-1(15),16,18-triene-17-carbaldehyde
SMILES C1COCCOC2=C(C=C(C=C2)C=O)OCCOCCO1

Spécifications

Melting Point 85°C
Quantity 1 g
Formula Weight 296.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Formylbenzo-15-crown 5-Ether