missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorostyrene, 97%, stabilized

Catalog No. AC168340010
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168340010 1 g Glass bottle
AC168340100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC168340010 Supplier Thermo Scientific Chemicals Supplier No. 168340010

CAS: 405-99-2 | C8H7F | 122.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 405-99-2 , 98-29-3
Molecular Formula C8H7F
Molecular Weight (g/mol) 122.14
MDL Number MFCD00000361
InChI Key JWVTWJNGILGLAT-UHFFFAOYSA-N
Synonym 4-fluorostyrene, 1-fluoro-4-vinylbenzene, p-fluorostyrene, benzene, 1-ethenyl-4-fluoro, styrene, p-fluoro, para-fluorostyrene, 1-fluoro-4-vinyl-benzene, 4-fluoro-styrene, acmc-1cszp
PubChem CID 67883
IUPAC Name 1-ethenyl-4-fluorobenzene
SMILES C=CC1=CC=C(C=C1)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0240g/mL
Boiling Point 145°C to 148°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5140 to 1.516
Linear Formula H2C=CHC6H4F
Quantity 1 g
Specific Gravity 1.024
Stabilizer 0.1% tert.-butylcatechol
Formula Weight 122.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Fluorostyrene, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-975-

RUO – Research Use Only