missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophthalonitrile, 98%

Catalog No. AAB2262403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2262403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2262403 Supplier Thermo Scientific Chemicals Supplier No. B2262403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65610-14-2 | C8H3FN2 | 146.124 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65610-14-2
Molecular Formula C8H3FN2
Molecular Weight (g/mol) 146.124
MDL Number MFCD00210430
InChI Key QQEKYCCJLSRLEC-UHFFFAOYSA-N
Synonym 4-fluorophthalonitrile, 1,2-benzenedicarbonitrile, 4-fluoro, 4-fluorophthalodinitrile, 1,2-dicyano-4-fluorobenzene, 4-fluoro-phthalonitrile, 2-cyano-5-fluorobenzonitrile, 4-fluorophthalonitrle, pubchem4027, acmc-1bb7s
PubChem CID 103423
IUPAC Name 4-fluorobenzene-1,2-dicarbonitrile
SMILES C1=CC(=C(C=C1F)C#N)C#N

Specifications

Melting Point 100°C to 102°C
Quantity 1 g
UN Number UN3439
Beilstein 2443236
Formula Weight 146.12
Percent Purity 98%
Chemical Name or Material 4-Fluorophthalonitrile
Hazard Category H300-H315-H319-H335
Hazard Statement GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only