missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophthalic Anhydride 98.0+%, TCI America™

Catalog No. F03225G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F03225G Supplier TCI America Supplier No. F03225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 319-03-9
Molecular Formula C8H3FO3
Molecular Weight (g/mol) 166.11
MDL Number MFCD00191363
InChI Key XVMKZAAFVWXIII-UHFFFAOYSA-N
Synonym 4-fluorophthalic anhydride, 5-fluoroisobenzofuran-1,3-dione, 5-fluoro-1,3-isobenzofurandione, 1,3-isobenzofurandione, 5-fluoro, 4-fluorophthalicanhydride, 4-fluorophthalic acid anhydride, 5-fluoro-1,3-dihydro-2-benzofuran-1,3-dione, 4-fluorobenzene-1,2-dicarboxylic anhydride, 5-fluoro-isobenzofuran-1,3-dione
PubChem CID 67572
IUPAC Name 5-fluoro-1,3-dihydro-2-benzofuran-1,3-dione
SMILES FC1=CC=C2C(=O)OC(=O)C2=C1

Specifications

Melting Point 79°C
Color White-Yellow
Boiling Point 206°C
Quantity 5 g
Formula Weight 166.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorophthalic Anhydride