missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylhydrazine Hydrochloride 97.0+%, TCI America™

Catalog No. F02465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02465G 5 g
F024625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02465G Supplier TCI America Supplier No. F02465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 823-85-8
Molecular Formula C6H8ClFN2
Molecular Weight (g/mol) 162.59
MDL Number MFCD00012942
InChI Key FEKUXLUOKFSMRO-UHFFFAOYSA-N
Synonym 4-fluorophenylhydrazine hydrochloride, 4-fluorophenyl hydrazine hydrochloride, 4-fluorophenylhydrazine hcl, 1-4-fluorophenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine hcl, p-fluorophenylhydrazine hydrochloride, 4-fluorphenyl hydrazine hydrochloride, 4-fluorophenyl hydrazine monohydrochloride, p-fluorophenylhydrazine hcl, hydrazine, 4-fluorophenyl-, monohydrochloride
PubChem CID 69981
IUPAC Name hydrogen (4-fluorophenyl)hydrazine chloride
SMILES [H+].[Cl-].NNC1=CC=C(F)C=C1

Specifications

Quantity 5 g
Formula Weight 162.59
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorophenylhydrazine Hydrochloride