missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylglyoxal hydrate, 98%, dry wt. basis

Catalog No. AAL2027206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2027206 5 g
AAL2027203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2027206 Supplier Thermo Scientific Chemicals Supplier No. L2027206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 403-32-7 | C8H5FO2 | 152.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 403-32-7
Molecular Formula C8H5FO2
Molecular Weight (g/mol) 152.12
MDL Number MFCD01733156
InChI Key IPWSCROFORAGJW-UHFFFAOYSA-N
Synonym 2-4-fluorophenyl-2-oxoacetaldehyde, 4-fluorophenylglyoxal, benzeneacetaldehyde, 4-fluoro-alpha-oxo, 4-fluoro-phenyl-oxo-acetaldehyde, 4-fluoro-alpha-oxobenzeneacetaldehyde, 4-fluorophenyl oxo acetaldehyde, p-fluorophenylglyoxal, p-fluorophenyl glyoxal, 4-fluorophenyl glyoxal, 1-4-fluorophenyl glyoxal
PubChem CID 101249
IUPAC Name 2-(4-fluorophenyl)-2-oxoacetaldehyde
SMILES FC1=CC=C(C=C1)C(=O)C=O

Specifications

Melting Point 84°C to 88°C
Quantity 5 g
Beilstein 2354778
Formula Weight 152.13 (Anhydrous)
Percent Purity 98%
Assay (dry wt.)
Chemical Name or Material 4-Fluorophenylglyoxal hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber CY1440000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only