missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylacetic acid, 98%

CAS: 405-50-5 | C8H7FO2 | 154.14 g/mol

Supplier:  Thermo Scientific Chemicals A1064614

 View more versions of this product

Catalog No. AAA1064614


Only null left
Explore available promotions

Description

Description

4-Fluorophenylacetic acid is used as an intermediate in the production of fluorinated anesthetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylacetic acid is used as an intermediate in the production of fluorinated anesthetics.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

4-Fluorophenylacetic acid
81°C to 85°C
>100°C (212°F)
C8H7FO2
25 g
14,4177
Insoluble in water.
C1=CC(=CC=C1CC(=O)O)F
154.14
154.14
405-50-5
164°C (2 mmHg)
Odorless
MFCD00004343
972145
4-fluorophenylacetic acid, 2-4-fluorophenyl acetic acid, p-fluorophenylacetic acid, 4-fluorophenyl acetic acid, benzeneacetic acid, 4-fluoro, 4-fluorobenzeneacetic acid, p-fluorophenyl acetic acid, acetic acid, p-fluorophenyl, unii-s5yjv7caz1, 4-fluoro-phenyl-acetic acid
MGKPFALCNDRSQD-UHFFFAOYSA-N
2-(4-fluorophenyl)acetic acid
9837
98%

Chemical Identifiers

405-50-5
154.14
MGKPFALCNDRSQD-UHFFFAOYSA-N
9837
C1=CC(=CC=C1CC(=O)O)F
C8H7FO2
MFCD00004343
4-fluorophenylacetic acid, 2-4-fluorophenyl acetic acid, p-fluorophenylacetic acid, 4-fluorophenyl acetic acid, benzeneacetic acid, 4-fluoro, 4-fluorobenzeneacetic acid, p-fluorophenyl acetic acid, acetic acid, p-fluorophenyl, unii-s5yjv7caz1, 4-fluoro-phenyl-acetic acid
2-(4-fluorophenyl)acetic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-972-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only