missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylacetic acid, 98%

Catalog No. p-7080911
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1064606 5 g
AAA1064614 25 g
AAA1064622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1064606 Supplier Thermo Scientific Chemicals Supplier No. A1064606
Only null left

CAS: 405-50-5 | C8H7FO2 | 154.14 g/mol

4-Fluorophenylacetic acid is used as an intermediate in the production of fluorinated anesthetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylacetic acid is used as an intermediate in the production of fluorinated anesthetics.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 405-50-5
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00004343
InChI Key MGKPFALCNDRSQD-UHFFFAOYSA-N
Synonym 4-fluorophenylacetic acid, 2-4-fluorophenyl acetic acid, p-fluorophenylacetic acid, 4-fluorophenyl acetic acid, benzeneacetic acid, 4-fluoro, 4-fluorobenzeneacetic acid, p-fluorophenyl acetic acid, acetic acid, p-fluorophenyl, unii-s5yjv7caz1, 4-fluoro-phenyl-acetic acid
PubChem CID 9837
IUPAC Name 2-(4-fluorophenyl)acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)F

Specifications

Melting Point 81°C to 85°C
Boiling Point 164°C (2 mmHg)
Flash Point >100°C (212°F)
Odor Odorless
Quantity 5 g
Beilstein 972145
Merck Index 14,4177
Solubility Information Insoluble in water.
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 4-Fluorophenylacetic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-972-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only