accessibility menu, dialog, popup

UserName

OHOF

CAS RN 405-50-5

IUPAC Name: 2-(4-fluorophenyl)acetic acid
Synonyms: P-fluorophenylacetic acid
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: MGKPFALCNDRSQD-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(F)C=C1
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)