missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl Methyl Sulfone 98.0+%, TCI America™

Catalog No. F06515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06515G 5 g
F065125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06515G Supplier TCI America Supplier No. F06515G
Only null left

Chemical Identifiers

CAS 455-15-2
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.189
MDL Number MFCD00039753
InChI Key DPJHZJGAGIWXTD-UHFFFAOYSA-N
Synonym 4-fluorophenyl methyl sulfone, 1-fluoro-4-methylsulfonyl benzene, benzene, 1-fluoro-4-methylsulfonyl, 1-fluoro-4-methylsulphonyl benzene, 1-fluoro-4-methanesulfonylbenzene, 4-fluorophenyl methyl sulphone, 4-fluorophenylmethylsulfone, sulfone, p-fluorophenyl methyl, methyl 4-fluorophenyl sulfone, 1-fluoro-4-methylsulfonyl-benzene
PubChem CID 67994
IUPAC Name 1-fluoro-4-methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=C(C=C1)F

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 5 g
Formula Weight 174.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorophenyl Methyl Sulfone