missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoronitrobenzene 98.0+%, TCI America™
Supplier: TCI America F0105100G
Specifications
| 4-Fluoronitrobenzene | |
| 207°C | |
| MFCD00007282 | |
| 2811 | |
| WFQDTOYDVUWQMS-UHFFFAOYSA-N | |
| 1-fluoro-4-nitrobenzene | |
| 9590 | |
| ≥98.0% (GC) |
| 350-46-9 | |
| C6H4FNO2 | |
| 100 g | |
| 4-fluoronitrobenzene, p-fluoronitrobenzene, p-nitrofluorobenzene, benzene, 1-fluoro-4-nitro, 4-nitrofluorobenzene, 1-nitro-4-fluorobenzene, 4-fluoro-1-nitrobenzene, 1-fluoro-4-nitro-benzene, unii-a2m2fh7xhh, p-fluoro nitrobenzene | |
| C1=CC(=CC=C1[N+](=O)[O-])F | |
| 141.101 | |
| 141.10 | |
| Crystalline Lumps |
Chemical Identifiers
| 350-46-9 | |
| 141.101 | |
| WFQDTOYDVUWQMS-UHFFFAOYSA-N | |
| 9590 | |
| C1=CC(=CC=C1[N+](=O)[O-])F |
| C6H4FNO2 | |
| MFCD00007282 | |
| 4-fluoronitrobenzene, p-fluoronitrobenzene, p-nitrofluorobenzene, benzene, 1-fluoro-4-nitro, 4-nitrofluorobenzene, 1-nitro-4-fluorobenzene, 4-fluoro-1-nitrobenzene, 1-fluoro-4-nitro-benzene, unii-a2m2fh7xhh, p-fluoro nitrobenzene | |
| 1-fluoro-4-nitrobenzene |
Safety and Handling
EINECSNumber : (3)-0441
RTECSNumber : DA1400000
TSCA : Yes