accessibility menu, dialog, popup

UserName

ONOF

CAS RN 350-46-9

IUPAC Name: 1-fluoro-4-nitrobenzene
Synonyms: 4-fluoronitrobenzene
Molecular Weight (g/mol): 141.1
Molecular Formula: C6H4FNO2
InChi Key: WFQDTOYDVUWQMS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C=C1
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)