missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoronitrobenzene 98.0+%, TCI America™

Catalog No. p-7129497
Change view
Click to view available options
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
F010525G 25 g
F0105100G 100 g
F0105500G 500 g
3 options

Catalog No. F010525G

Supplier: TCI America F010525G

Only null left

Chemical Identifiers

CAS 350-46-9
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.101
MDL Number MFCD00007282
InChI Key WFQDTOYDVUWQMS-UHFFFAOYSA-N
Synonym 4-fluoronitrobenzene, p-fluoronitrobenzene, p-nitrofluorobenzene, benzene, 1-fluoro-4-nitro, 4-nitrofluorobenzene, 1-nitro-4-fluorobenzene, 4-fluoro-1-nitrobenzene, 1-fluoro-4-nitro-benzene, unii-a2m2fh7xhh, p-fluoro nitrobenzene
PubChem CID 9590
IUPAC Name 1-fluoro-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])F

Specifications

Boiling Point 207°C
Quantity 25 g
UN Number 2811
Formula Weight 141.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Fluoronitrobenzene
EINECSNumber (3)-0441
RTECSNumber DA1400000
TSCA Yes