missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoroindazole 98.0+%, TCI America™

Catalog No. F10371G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10371G Supplier TCI America Supplier No. F10371G
Only null left

Chemical Identifiers

CAS 341-23-1
Molecular Formula C7H5FN2
Molecular Weight (g/mol) 136.129
MDL Number MFCD07371561
InChI Key YTNDJUBXSVSRQK-UHFFFAOYSA-N
PubChem CID 22477757
IUPAC Name 4-fluoro-1H-indazole
SMILES C1=CC2=C(C=NN2)C(=C1)F

Specifications

Melting Point 132°C
Color Yellow
Quantity 1 g
Formula Weight 136.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoroindazole