missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorocinnamaldehyde 95.0+%, TCI America™

Catalog No. F07225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07225G 5 g
F072225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07225G Supplier TCI America Supplier No. F07225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24654-55-5
Molecular Formula C9H7FO
Molecular Weight (g/mol) 150.152
MDL Number MFCD03425895
InChI Key YSIYEWBILJZDQH-OWOJBTEDSA-N
Synonym 3-(4-Fluorophenyl)acrylaldehyde, 3-(4-Fluorophenyl)-2-propenal
PubChem CID 11217342
IUPAC Name (E)-3-(4-fluorophenyl)prop-2-enal
SMILES C1=CC(=CC=C1C=CC=O)F

Specifications

Color Yellow
Boiling Point 95°C
Quantity 5 g
Formula Weight 150.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorocinnamaldehyde
TSCA No
Recommended Storage Refrigerator