missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzotrifluoride 98.0+%, TCI America™

Catalog No. F03575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03575G 5 g
F035725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03575G Supplier TCI America Supplier No. F03575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 402-44-8
Molecular Formula C7H4F4
Molecular Weight (g/mol) 164.103
MDL Number MFCD00000399
InChI Key UNNNAIWPDLRVRN-UHFFFAOYSA-N
Synonym 4-fluorobenzotrifluoride, 1-fluoro-4-trifluoromethyl benzene, p-fluorobenzotrifluoride, benzene, 1-fluoro-4-trifluoromethyl, alpha,alpha,alpha,4-tetrafluorotoluene, p-fluorotrifluoromethylbenzene, p-trifluoromethyl fluorobenzene, 4-flourobenzotrifluoride, a,a,a,4-tetrafluorotoluene, 4-trifluoromethyl fluorobenzene
PubChem CID 67873
IUPAC Name 1-fluoro-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)F

Specifications

Color Colorless
Boiling Point 103°C
Quantity 5 g
UN Number 1993
Formula Weight 164.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzotrifluoride
TSCA Yes
Recommended Storage Refrigerator