missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzophenone 99.0+%, TCI America™

Catalog No. F01615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F01615G 5 g
F016125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F01615G Supplier TCI America Supplier No. F01615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 345-83-5
Molecular Formula C13H9FO
Molecular Weight (g/mol) 200.21
MDL Number MFCD00000352
InChI Key OGTSHGYHILFRHD-UHFFFAOYSA-N
Synonym 4-fluorobenzophenone, 4-fluorophenyl phenyl methanone, p-fluorobenzophenone, methanone, 4-fluorophenyl phenyl, 4-fluorophenyl phenyl ketone, 4-fluorophenyl-phenylmethanone, 4'-fluorobenzophenone, 4-fluoro-benzophenone, pubchem3308, benzophenone, 4-fluoro
PubChem CID 67663
IUPAC Name (4-fluorophenyl)(phenyl)methanone
SMILES FC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 49°C
Color White
Boiling Point 161°C
Quantity 5 g
Formula Weight 200.21
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorobenzophenone