missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzaldehyde, 98%

Catalog No. AAA1538314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1538314 25 g
AAA1538322 100 g
AAA1538336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1538314 Supplier Thermo Scientific Chemicals Supplier No. A1538314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 459-57-4 | C7H5FO | 124.114 g/mol

4-Fluorobenzaldehyde is used in the preparation of pyrazolopyridine derivative, which finds application as mitogen-activated protein kinase (MAPK) inhibitor. It also serves as a synthetic intermediate in the preparation of pharmaceutical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzaldehyde is used in the preparation of pyrazolopyridine derivative, which finds application as mitogen-activated protein kinase (MAPK) inhibitor. It also serves as a synthetic intermediate in the preparation of pharmaceutical compounds.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.

Chemical Identifiers

CAS 459-57-4
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.114
MDL Number MFCD00003378
InChI Key UOQXIWFBQSVDPP-UHFFFAOYSA-N
Synonym p-fluorobenzaldehyde, benzaldehyde, 4-fluoro, 4-fluoro-benzaldehyde, para-fluorobenzaldehyde, p-fluorbenzaldehyde, fluorobenzaldehyde 4-, benzaldehyde, p-fluoro, pfad, 4-fluoro benzaldehyde, para fluoro benzaldehyde
PubChem CID 68023
IUPAC Name 4-fluorobenzaldehyde
SMILES C1=CC(=CC=C1C=O)F

Specifications

Melting Point -10°C
Density 1.175
Boiling Point 180°C to 181°C
Flash Point 56°C (133°F)
Refractive Index 1.521
Quantity 25 g
UN Number UN1989
Beilstein 385857
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 124.11
Percent Purity 98%
Chemical Name or Material 4-Fluorobenzaldehyde
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber 207-293-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only