missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Fluoroacétophénone 98,0+%, TCI America™
Spécifications
Specifications
| Color | Jaune |
| Boiling Point | 77°C |
| Quantity | 25 g |
| Formula Weight | 138.14 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 4′-Fluoroacetophenone |
Safety and Handling
| TSCA | Yes |