missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-o-phenylenediamine, 97%

Catalog No. AAA1397706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1397706 5 g
AAA1397714 25 g
AAA1397722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1397706 Supplier Thermo Scientific Chemicals Supplier No. A1397706
Only null left

CAS: 367-31-7 | C6H7FN2 | 126.134 g/mol

4-Fluoro-o-phenylenediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-o-phenylenediamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 367-31-7
Molecular Formula C6H7FN2
Molecular Weight (g/mol) 126.134
MDL Number MFCD00042228
InChI Key KWEWNOOZQVJONF-UHFFFAOYSA-N
Synonym 1,2-diamino-4-fluorobenzene, 4-fluoro-1,2-phenylenediamine, 3,4-diaminofluorobenzene, 4-fluoro-o-phenylenediamine, 2-amino-4-fluoroaniline, 4-fluoro-1,2-diaminobenzene, 3,4-diamino-1-fluorobenzene, 5-fluoro-1,2-diaminobenzene, 4-fluoro-1,2-benzenediamine, 1,2-benzenediamine, 4-fluoro
PubChem CID 164584
IUPAC Name 4-fluorobenzene-1,2-diamine
SMILES C1=CC(=C(C=C1F)N)N

Specifications

Melting Point 91°C to 95°C
Quantity 5 g
UN Number UN2811
Beilstein 2081075
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 126.13
Percent Purity 97%
Chemical Name or Material 4-Fluoro-o-phenylenediamine
Hazard Category H302+H332-H311-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 206-691-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only