missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-DL-phenylalanine, 98+%

Catalog No. AAL0758506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0758506 5 g
AAL0758503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0758506 Supplier Thermo Scientific Chemicals Supplier No. L0758506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51-65-0 | C9H10FNO2 | 183.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-65-0
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.18
MDL Number MFCD00002600,MFCD00063064
InChI Key XWHHYOYVRVGJJY-UHFFFAOYNA-N
Synonym 4-fluoro-dl-phenylalanine, 4-fluorophenylalanine, p-fluorophenylalanine, 2-amino-3-4-fluorophenyl propanoic acid, dl-p-fluorophenylalanine, p-fluoro-dl-phenylalanine, alnasid, dl-4-fluorophenylalanine, dl-4-f-phe-oh, h-dl-phe 4-f-oh
PubChem CID 4654
ChEBI CHEBI:84060
IUPAC Name 2-amino-3-(4-fluorophenyl)propanoic acid
SMILES NC(CC1=CC=C(F)C=C1)C(O)=O

Specifications

Melting Point ∽254°C (decomposition)
Beilstein 29338793
Quantity 5 g
Formula Weight 183.18
Percent Purity ≥98%
Chemical Name or Material 4-Fluoro-DL-phenylalanine
EINECSNumber 200-113-7
RTECSNumber AY6315000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only