missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-alpha-toluenesulfonyl chloride, 97%

Catalog No. AAH31605MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31605MD 250 mg
AAH3160503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH31605MD Supplier Thermo Scientific Chemicals Supplier No. H31605MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 103360-04-9 | C7H6ClFO2S | 208.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103360-04-9
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.631
MDL Number MFCD01631930
InChI Key UUQGWVIRPCRTSA-UHFFFAOYSA-N
Synonym 4-fluorophenyl methanesulfonyl chloride, 4-fluorobenzylsulfonyl chloride, 4-fluoro-phenyl-methanesulfonyl chloride, 4-fluorophenyl methyl sulfonyl chloride, chloro 4-fluorophenyl methyl sulfone, 4-fluorophenyl methanesulphonyl chloride, 4-fluoro-alpha-toluenesulfonyl chloride, benzenemethanesulfonyl chloride, 4-fluoro, 4-fluoro-phenyl-methane sulfonyl chloride, pubchem10056
PubChem CID 2759108
IUPAC Name (4-fluorophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC=C1CS(=O)(=O)Cl)F

Specifications

Melting Point 58°C to 61°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 208.64
Percent Purity 97%
Chemical Name or Material 4-Fluoro-alpha-toluenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only