missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methoxybenzoic acid, 95%, Thermo Scientific Chemicals

Catalog No. AAL1967306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1967306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1967306 Supplier Thermo Scientific Chemicals Supplier No. L1967306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 82846-18-2 | Molecular Formula: C8H7FO3 | Molecular Weight: 170.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82846-18-2
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00272134
InChI Key LWGCZCMLPRMKIZ-UHFFFAOYSA-N
Synonym 3-methoxy-4-fluorobenzoic acid, 4-fluoro-m-anisic acid, benzoic acid, 4-fluoro-3-methoxy, 4-fluoro-3-methoxybenzoicacid, 4-fluoro-3-methoxy-benzoic acid, 4-fluoranyl-3-methoxy-benzoic acid, pubchem1394, acmc-209pqm, ksc495c0r, rarechem al bo 0973
PubChem CID 598436
IUPAC Name 4-fluoro-3-methoxybenzoic acid
SMILES COC1=C(C=CC(=C1)C(=O)O)F

Specifications

Melting Point 206°C to 208°C
Quantity 5 g
Beilstein 4178071
Formula Weight 170.14
Percent Purity 95%
Chemical Name or Material 4-Fluoro-3-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only