missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitrophenol 98.0+%, TCI America™

Numéro de catalogue. p-7129641
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
F03675G 5 g
F036725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. F03675G Fournisseur TCI America Code fournisseur F03675G
Il en reste null

Identifiants chimiques

CAS 394-33-2
Molecular Formula C6H4FNO3
Molecular Weight (g/mol) 157.1
MDL Number MFCD00042528
InChI Key ZHRLVDHMIJDWSS-UHFFFAOYSA-N
Synonym 2-nitro-4-fluorophenol, phenol, 4-fluoro-2-nitro, phenol,2-nitro,4-fluoro, 4-fluoro-2-nitro-phenol, 4-fluoronitrophenol, pubchem1503, acmc-1ah4y, ksc493s6p, 5-fluoro-2-hydroxynitrobenzene, 4-fluoro-2-nitrophenol
PubChem CID 136236
IUPAC Name 4-fluoro-2-nitrophenol
SMILES C1=CC(=C(C=C1F)[N+](=O)[O-])O

Spécifications

Melting Point 75°C
Quantity 5 g
Formula Weight 157.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-nitrophenol