missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog No. AAH6205903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6205903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6205903 Supplier Thermo Scientific Chemicals Supplier No. H6205903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1288978-82-4 | Molecular Formula: C12H15BFNO4 | Molecular Weight: 267.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1288978-82-4
Molecular Formula C12H15BFNO4
Molecular Weight (g/mol) 267.06
MDL Number MFCD16996295
InChI Key UEMDKWBOUAQJJU-UHFFFAOYSA-N
PubChem CID 71744276
IUPAC Name 2-(4-fluoro-2-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=C(C=C(F)C=C1)[N+]([O-])=O

Specifications

Quantity 1 g
Synonym 2-4-fluoro-2-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-fluoro-2-nitrobenzeneboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-4-fluoro-2-nitrophenyl-4,4,5,5-tetramethyl, 1,3,2-dioxaborolane,2-4-fluoro-2-nitrophenyl-4,4,5,5-tetramethyl, 4-fluoro-2-nitrophenylboronic acid pinacol ester, 4-fluoro-2-nitrophenyl boronic acid pinacol ester
Formula Weight 267.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Fluoro-2-nitrobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only