missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitroanisole, 98%

Catalog No. AAH6111014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6111014 25 g
AAH6111006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6111014 Supplier Thermo Scientific Chemicals Supplier No. H6111014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 445-83-0 | C7H6FNO3 | 171.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 445-83-0
Molecular Formula C7H6FNO3
Molecular Weight (g/mol) 171.13
MDL Number MFCD00013375
InChI Key FWLPYISRFBKEKV-UHFFFAOYSA-N
Synonym 4-fluoro-2-nitroanisole, 4-fluoro-2-nitroanisol, benzene, 4-fluoro-1-methoxy-2-nitro, 4-fluoro-1-methoxy-2-nitro-benzene, 5-fluoro-2-methoxynitrobenzene, 4-fluor-1-methoxy-2-nitrobenzol, pubchem1966, acmc-1amcp, 4-fluoro-2-nitro-anisole, ksc235o8j
PubChem CID 565656
IUPAC Name 4-fluoro-1-methoxy-2-nitrobenzene
SMILES COC1=C(C=C(F)C=C1)[N+]([O-])=O

Specifications

Melting Point 62°C to 64°C
Color Brown-Yellow
Quantity 25 g
Formula Weight 171.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Fluoro-2-nitroanisole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only