missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitroaniline, 98%

Catalog No. AAA1068518
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1068518 50 g
AAA1068522 100 g
AAA1068536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1068518 Supplier Thermo Scientific Chemicals Supplier No. A1068518
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 364-78-3 | C6H5FN2O2 | 156.12 g/mol

4-Fluoro-2-nitroaniline is used as a reagent in the preparation of 4-fluoro-o-phenylenediamine, 4-fluoro-N-ethyl-2-nitroaniline and N-(4-fluoro-2-nitrophenyl)-beta-alanine. It is also employed as a pharmaceutical intermediate. It acts as monodendate O-bonded ligand used in the coordination chemistry to form complexes with copper(II), nickel(II) and cobalt(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-2-nitroaniline is used as a reagent in the preparation of 4-fluoro-o-phenylenediamine, 4-fluoro-N-ethyl-2-nitroaniline and N-(4-fluoro-2-nitrophenyl)-beta-alanine. It is also employed as a pharmaceutical intermediate. It acts as monodendate O-bonded ligand used in the coordination chemistry to form complexes with copper(II), nickel(II) and cobalt(II).

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and strong acids.

Chemical Identifiers

CAS 364-78-3
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00007830
InChI Key PUGDHSSOXPHLPT-UHFFFAOYSA-N
Synonym 2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421
PubChem CID 67769
IUPAC Name 4-fluoro-2-nitroaniline
SMILES NC1=CC=C(F)C=C1[N+]([O-])=O

Specifications

Melting Point 90°C to 94°C
Flash Point 89°C (192°F)
Quantity 50 g
Beilstein 2210197
Solubility Information Insoluble in water.
Formula Weight 156.12
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-nitroaniline
Hazard Category H228-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 206-666-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only