missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-2'-hydroxyacetophenone 97.0+%, TCI America™

Catalog No. F07235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07235G 5 g
F072325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07235G Supplier TCI America Supplier No. F07235G
Only null left

Chemical Identifiers

CAS 1481-27-2
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00143203
InChI Key HLTBTUXAMVOKIH-UHFFFAOYSA-N
Synonym 4'-fluoro-2'-hydroxyacetophenone, 1-4-fluoro-2-hydroxyphenyl ethanone, 4-fluoro-2-hydroxyacetophenone, 1-4-fluoro-2-hydroxyphenyl ethan-1-one, ethanone, 1-4-fluoro-2-hydroxyphenyl, 1-4-fluoro-2-hydroxy-phenyl ethanone, 1-acetyl-4-fluoro-2-hydroxybenzene, pubchem13252, acmc-1bzss, 4'-fluoro-2'-hydroxy
PubChem CID 2737326
IUPAC Name 1-(4-fluoro-2-hydroxyphenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(F)C=C1O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4′-Fluoro-2′-hydroxyacetophenone