missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-1,3-dioxolan-2-one, 98%

Catalog No. AAH6150222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6150222 100 g
AAH6150214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6150222 Supplier Thermo Scientific Chemicals Supplier No. H6150222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 114435-02-8 | C3H3FO3 | 106.052 g/mol

4-Fluoro-1,3-dioxolan-2-one is used as an electrolyte additive in lithium ion batteries. It also enables the formation of thin, smooth and stable passive solid electrolyte interphase (SEI) layer, which increases the cycling efficiency and discharge capacity retention of the secondary battery. Further, it is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-1,3-dioxolan-2-one is used as an electrolyte additive in lithium ion batteries. It also enables the formation of thin, smooth and stable passive solid electrolyte interphase (SEI) layer, which increases the cycling efficiency and discharge capacity retention of the secondary battery. Further, it is used as an intermediate in organic synthesis.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 114435-02-8
Molecular Formula C3H3FO3
Molecular Weight (g/mol) 106.052
MDL Number MFCD06247543
InChI Key SBLRHMKNNHXPHG-UHFFFAOYSA-N
Synonym fluoroethylene carbonate, 1,3-dioxolan-2-one, 4-fluoro, fluoro ethylene carbonate, 4-fluoro-1,3-dioxalan-2-one, 4-fluoro-2-oxo-1,3-dioxolane, 4-fluoro-ethylene carbonate, 4-fluoro-ethylene carboante, ksc219e1h, fec, lithium ion battery grade
PubChem CID 2769656
IUPAC Name 4-fluoro-1,3-dioxolan-2-one
SMILES C1C(OC(=O)O1)F

Specifications

Melting Point 18°C to 23°C
Color Colorless
Boiling Point 212°C
Flash Point >102°C (216°F)
Quantity 100 g
Solubility Information Slightly miscible with water.
Formula Weight 106.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-1,3-dioxolan-2-one
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 483-360-5
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only