missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyltoluene 97.0+%, TCI America™

Catalog No. p-7128885
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E018625ML 25 mL
E0186100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E018625ML Supplier TCI America Supplier No. E018625ML

Chemical Identifiers

CAS 622-96-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00009263
InChI Key JRLPEMVDPFPYPJ-UHFFFAOYSA-N
Synonym 4-ethyltoluene, p-ethyltoluene, benzene, 1-ethyl-4-methyl, 4-methylethylbenzene, toluene, p-ethyl, p-ethylmethylbenzene, p-methylethylbenzene, 1-methyl-4-ethylbenzene, 1-ethyl-4-methyl-benzene, para-ethyltoluene
PubChem CID 12160
IUPAC Name 1-ethyl-4-methylbenzene
SMILES CCC1=CC=C(C=C1)C

Specifications

Color Colorless
Boiling Point 161°C
Quantity 25 mL
UN Number 3295
Formula Weight 120.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethyltoluene
EINECSNumber (3)-0015
RTECSNumber XT2550000
TSCA Yes