missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethylsulfonyl)benzeneboronic acid, 98+%

Catalog No. AAL17814MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17814MD 250 mg
AAL1781403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL17814MD Supplier Thermo Scientific Chemicals Supplier No. L17814MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 352530-24-6 | C8H11BO4S | 214.042 g/mol

It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 352530-24-6
Molecular Formula C8H11BO4S
Molecular Weight (g/mol) 214.042
MDL Number MFCD02093076
InChI Key CVBNDDGEVPNUNA-UHFFFAOYSA-N
Synonym 4-ethylsulfonyl phenylboronic acid, 4-ethylsulfonyl phenyl boronic acid, 4-ethanesulfonyl benzeneboronic acid, 4-ethanesulfonyl phenylboronic acid, 4-ethylsulfonylphenyl boronic acid, 4-ethylsulphonyl benzeneboronic acid, boronic acid, 4-ethylsulfonyl phenyl, 4-ethanesulfonyl phenyl boranediol, 4-ethanesulphonyl benzeneboronic acid
PubChem CID 2773420
IUPAC Name (4-ethylsulfonylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)S(=O)(=O)CC)(O)O

Specifications

Melting Point 135°C to 139°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 214.05
Percent Purity ≥98%
Chemical Name or Material 4-(Ethylsulfonyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only