missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethoxycarbonyl)butylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58484AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58484AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58484AD Supplier Thermo Scientific Chemicals Supplier No. H58484AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 131379-16-3 | C9H9IO2Zn | 341.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Ethoxycarbonyl)phenylzinc iodide is used as pharmaceutical intermediates.

Solubility
Reacts with water.

Notes
Store at 4°C. Air Sensitive. Store under dry inert gas. Store away from air. Inco

Chemical Identifiers

CAS 131379-16-3
Molecular Formula C9H9IO2Zn
Molecular Weight (g/mol) 341.45
MDL Number MFCD00671982
InChI Key DVAYVJGYZRYJEV-UHFFFAOYSA-M
Synonym 4-ethoxycarbonylphenylzinc iodide, 4-ethoxycarbonyl phenylzinc iodide, 4-ethoxycarbonyl phenyl iodo zinc, 4-ethoxycarbonyl phenyl zinc ii iodide, 4-ethoxycarbonyl phenylzinc iodide solution, 4-ethoxycarbonylphenylzinc iodide 0.5 m in tetrahydrofuran, 4-ethoxycarbonylphenylzinc iodide, 0.5m in tetrahydrofuran, 4-ethoxycarbonyl phenylzinc iodide solution, 0.5 m in thf, 4-ethoxycarbonyl phenylzinc iodide, 0.5m in thf, packaged under argon in resealable chemseal bottles
PubChem CID 5107659
IUPAC Name 4-(ethoxycarbonyl)benzen-1-ide; iodozincylium
SMILES [Zn+]I.CCOC(=O)C1=CC=[C-]C=C1

Specifications

Density 1.023
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Solubility Information Reacts with water.
Formula Weight 341.45
Chemical Name or Material 4-(Ethoxycarbonyl)phenylzinc iodide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H260-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only