missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethoxycarbonyl)-3-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. E12071G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E12071G Supplier TCI America Supplier No. E12071G

Specifications

Chemical Name or Material 4-(Ethoxycarbonyl)-3-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 874288-38-7
Melting Point 161°C
Color White
Molecular Formula C9H10BFO4
Quantity 1 g
MDL Number MFCD06656274
Synonym 4-ethoxycarbonyl-3-fluorophenylboronic acid, 4-ethoxycarbonyl-3-fluorobenzeneboronic acid, ethyl 4-borono-2-fluorobenzoate, 4-ethoxycarbonyl-3-fluorophenyl boronic acid, 4-ethoxycarbonyl-3-fluoro-phenyl boronic acid, 4-ethoxycarbonyl-3-fluorophenylboronicacid, 3-fluoro-4-ethoxycarbonylphenylboronic acid, 4-borono-2-fluoro-benzoic acid, 1-ethyl ester, benzoic acid, 4-borono-2-fluoro-, 1-ethyl ester, acmc-209qm0
InChI Key CXSPPZZTTSHTMP-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)C(=O)OCC)F)(O)O
IUPAC Name (4-ethoxycarbonyl-3-fluorophenyl)boronic acid
Molecular Weight (g/mol) 211.983
PubChem CID 2763245
Formula Weight 211.98
Physical Form Crystalline Powder
Show More Show Less